Molybden 1.000 mg/l i vann AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86692.260EA 520 NOK
86692.260 86692.180
Molybden 1.000 mg/l i vann AVS TITRINORM standard til AAS
Standarder Kationstandarder Molybdenum Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%