Neodymium 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86695.180EA 510 NOK
86695.180 86695.260
Neodymium 1.000 mg/l i fortynnet salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Neodymium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%