Rhenium 1.000 mg/l Re in dil. salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86703.180EA 1120 NOK
86703.180 86703.260
Rhenium 1.000 mg/l Re in dil. salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Rhenium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%