Svovel 1.000 mg/l i vann AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86706.180EA 238 NOK
86706.180 86706.260
Svovel 1.000 mg/l i vann AVS TITRINORM standard til AAS
Standarder Kationstandarder Sulphur Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%