Antimon 1.000 mg/l Sb in 5% salpetersyre with hydrofluoric acid (max. 1%) AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86707.260EA 470 NOK
86707.260 86707.180
Antimon 1.000 mg/l Sb in 5% salpetersyre with hydrofluoric acid (max. 1%) AVS TITRINORM standard til AAS
Standarder Kationstandarder Antimony Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%