Tantalum 1.000 mg/l Ta in 5% salpetersyre with hydrofluoric acid (max. 1%) AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86714.180EA 294 NOK
86714.180 86714.260
Tantalum 1.000 mg/l Ta in 5% salpetersyre with hydrofluoric acid (max. 1%) AVS TITRINORM standard til AAS
Standarder Kationstandarder Tantalum Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%