Thulium 1.000 mg/l Tm in dil. salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86719.180EA 294 NOK
86719.180 86719.260
Thulium 1.000 mg/l Tm in dil. salpetersyre AVS TITRINORM standard til AAS
Standarder Kationstandarder Thulium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%