Wolfram 1.000 mg/l W in salpetersyre with hydrofluoric acid (each max. 1%) AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
86722.260EA 577.5 NOK
86722.260 86722.180
Wolfram 1.000 mg/l W in salpetersyre with hydrofluoric acid (each max. 1%) AVS TITRINORM standard til AAS
Standarder Kationstandarder Tungsten Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%