Tetrabutylammonium dihydrogen phosphate Ionpair, Supelco®

Leverandør: Merck

Synonymer: Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate

86899-6X1AMP-F
86899-6X1AMP-FEA 4600 NOK
86899-6X1AMP-F
Tetrabutylammonium dihydrogen phosphate Ionpair, Supelco®
Tetrabutylammonium dihydrogen phosphate
Formel: C₁₆H₃₈NO₄P
MW: 339.46 g/mol
MDL nummer: MFCD00064526
CAS-nummer: 5574-97-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a089b25147c132023566d34125908af66981b3c3&vid=c2c5ca6a9dddbb17380fef4452042077f906bdce&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR