VWR® ARISTAR®, Multi-Element Standards, Cationic for Ion Chromatography
Leverandør: VWR Chemicals
87041.180EA 1410 NOK
87041.180 87042.180
VWR® ARISTAR®, Multi-Element Standards, Cationic for Ion Chromatography
Standarder Kationstandarder Multi-Element Standards
  • Traceable to NIST
  • Manufactured and tested in ISO Guide 34 / ISO17025 accredited facilities
  • Supplied with a Certificate of Analysis
Ion chromatography cation multi-element standard solution VII, ARISTAR® (100 mg/L: Li, Na, K, NH₄, Mn, Ca, Mg, Sr, Ba in 0,1% HNO₃)