Glykolsyre ≥99.5% (by acidimetry), AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Danger

Synonymer: 2-Hydroxy acid, 2-Hydroxyacetic acid

87061.180EA 975 NOK
87061.180
Glykolsyre ≥99.5% (by acidimetry), AnalaR NORMAPUR® analytical reagent
Glykolsyre
Formel: C₂H₄O₃
MW: 76.05 g/mol
Kokepunkt: 266
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004312
CAS-nummer: 79-14-1
FN: 3261
ADR: 8 II
Merck Indeks: 13,04511

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white crystals
Assay (acidimetric) min. 99.5 %
Water max. 0.5 %
Melting range (lower value) min. 75 °C
Melting range (upper value) max. 79 °C
Iron (Fe) max. 5 ppm
Heavy metals (as Pb) max. 10 ppm
Identity (IR) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR