Perchloric acid 0.01 mol/l (0.01 N) anhydrous acetic acid

Leverandør: VWR Chemicals
Danger

87116.290EA 444 NOK
87116.290
Perchloric acid 0.01 mol/l (0.01 N) anhydrous acetic acid
Perchloric acid
Formel: HClO₄
MW: 100.46 g/mol
Kokepunkt: 39
Smeltepunkt: -101
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
EINECS: 231-512-4
FN: 2789
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR