Tetrahydrofuran-D8 (99.5% D) for NMR spectroscopy

Leverandør: VWR Chemicals
Danger

Synonymer: Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8

87158.0010EA 5675 NOK
87158.0010
Tetrahydrofuran-D8 (99.5% D) for NMR spectroscopy
Tetrahydrofuran-D8
Formel: C₄D₈O
MW: 80.04 g/mol
Kokepunkt: 65
Smeltepunkt: –108 °C
Tetthet: 0.98 g/cm³ (20 °C)
Flash punkt: –17 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00044238
CAS-nummer: 1693-74-9
EINECS: 216-898-4
FN: 2056
ADR: 3 II

Bestill nå

Spesifikasjon Testresultater

Assay (on anhydrous substance) Min. 99.9 %
Isotopic enrichment (FT NMR 400 MHz)(D) Min. 99.50 %
Water (HDO+D2O) Max. 0.0500 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR