Tetradekylbensen, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

87204-500MG
87204-500MGEA 1900 NOK
87204-500MG
Tetradekylbensen, Supelco®
Tetradekylbensen
Formel: C₂₀H₃₄
MW: 274,49 g/mol
Kokepunkt: 354 °C (1013 hPa)
Smeltepunkt: 16,1 °C
Tetthet: 0,8559 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00015080
CAS-nummer: 1459-10-5
EINECS: 215-950-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9324e2812421d4571f80b03ee8d65a4d91213205&vid=de2e0784e4c0e07b06a948e229beae430ff989fc&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR