Tetrahexylammonium hydrogen sulphate, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

87299-25G-F 87299-5G-F
87299-25G-FEA 9080 NOK
87299-25G-F 87299-5G-F
Tetrahexylammonium hydrogen sulphate, Supelco®
Tetrahexylammonium hydrogen sulphate
Formel: C₂₄H₅₃NO₄S
MDL nummer: MFCD00037675
CAS-nummer: 32503-34-7
EINECS: 251-069-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e34d4223ffefe1be4ddaf8d87e9efe8b42f85777&vid=60e233cf860a77808d1b0dc36b768c1e5c2f5f87&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR