Natriumtetrahydroborat 4.4 M vandig løsning stable

Leverandør: Thermo Fisher Scientific
Danger

87659.AE 87659.AP 87659.K3
87659.AEEA 495 NOK
87659.AE 87659.AP 87659.K3
Natriumtetrahydroborat 4.4 M vandig løsning stable
Sodium borohydride

CAS No.: 16940-66-2 Molecular Formula: BH4Na Formula Weight: 37.83 Physical Form: Liquid Density: 1.4 Sensitivity: Air Sensitive Appearance: Colourless UN No.: UN3320 MDL No.: MFCD00003518

Formel: NaBH₄
Tetthet: 1,4
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003518
CAS-nummer: 16940-66-2
EINECS: 241-004-4
FN: 3320
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR