Saltsyre 1% Reag. Ph. Eur.

Leverandør: VWR Chemicals
Warning

87684.290EA 2075 NOK
87684.290
Saltsyre 1% Reag. Ph. Eur.
Saltsyre
Formel: HCl
Kokepunkt: >100 °C (1013 hPa)
Tetthet: 1,02…1.05 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR