Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l vandig løsning Reag. Ph. Eur. 1038100 redox indicator

Leverandør: VWR Chemicals

87702.180EA 1850 NOK
87702.180
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 mol/l vandig løsning Reag. Ph. Eur. 1038100 redox indicator
Ferroin (1,10-phenanthrolineferrous sulphate complex)

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: C₃₆H₂₄FeN₆SO₄
Tetthet: 1,01 g/cm³ (20 °C)
MDL nummer: MFCD00036428
CAS-nummer: 14634-91-4
EINECS: 238-676-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR