Titanium trichloride sulphuric acid reagent Reag. Ph. Eur. 1091202
Leverandør: VWR Chemicals
Danger
87711.180EA 1035 NOK
87711.180
Titanium trichloride sulphuric acid reagent Reag. Ph. Eur. 1091202
Titanium trichloride sulphuric acid reagent
Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
MW: 240.01 g/mol
Lagringstemperatur: Romtemperatur
FN: 2796
ADR: 8 II