Tetramethylammonium hydrogen sulphate, Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

87724-10G-F 87724-50G-F
87724-10G-FEA 1825 NOK
87724-10G-F 87724-50G-F
Tetramethylammonium hydrogen sulphate, Supelco®
Tetramethylammonium hydrogen sulphate
Formel: C₄H₁₃NO₄S
MDL nummer: MFCD00036149
CAS-nummer: 80526-82-5
EINECS: 279-490-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4651ae55de3053434799889a9abbcd8f4784d91d&vid=e48768f22880db16368900698207f25ca9c6cb9e&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR