BRP Indicator solution Reag. Ph. Eur. 1013000
Leverandør: VWR Chemicals
87799.180EA 755 NOK
87799.180
BRP Indicator solution Reag. Ph. Eur. 1013000
BRP Indicator solution
Solution of Bromothymol Blue R, Methyl Red R and Phenolphtthalein R in alcohol