Kalsiumklorid 0.01 mol/l (0.02 N) aqueous solution Reag. Ph. Eur. 1014602

Leverandør: VWR Chemicals

Synonymer: Chloro calcium, Calcium dichloride, CaCl2

87801.290EA 1455 NOK
87801.290
Kalsiumklorid 0.01 mol/l (0.02 N) aqueous solution Reag. Ph. Eur. 1014602
Kalsiumklorid

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: CaCl₂
MW: 110.98 g/mol
Kokepunkt: 1600
Smeltepunkt: 772
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010903
CAS-nummer: 10043-52-4
EINECS: 233-140-8
Merck Indeks: 14,01659

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR