1,2-Dinitrobensen Reag. Ph. Eur. 1031201

Leverandør: VWR Chemicals

87818.290EA 3325 NOK
87818.290
1,2-Dinitrobensen Reag. Ph. Eur. 1031201
1,2-Dinitrobensen
Formel: C₆H₄N₂O₄
MW: 168.11 g/mol
Kokepunkt: 297
MDL nummer: MFCD00007093
CAS-nummer: 528-29-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR