Difenylamin 1% Reag. Ph. Eur. 1032102

Leverandør: VWR Chemicals
Danger

Synonymer: N-Phenylaniline, N-Phenylbenzeneamine

87823.290EA 2175 NOK
87823.290
Difenylamin 1% Reag. Ph. Eur. 1032102
Difenylamin

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: (C₆H₅)₂NH
MW: 169.23 g/mol
Kokepunkt: 302
Flash punkt: 153
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003014
CAS-nummer: 122-39-4
EINECS: 204-539-4
FN: 1830
ADR: 8 II
Merck Indeks: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR