Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702

Leverandør: VWR Chemicals

Synonymer: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate

87830.290EA 3625 NOK
87830.290
Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702
Ammonium iron(II) sulphate

Other name: Ferric ammonium sulphate solution R2


Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: H₈FeN₂O₈S₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00051039
CAS-nummer: 10045-89-3
FN: 3264
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR