Formaldehyd 35% in aqueous solution stabilised Reag. Ph. Eur. 1039101
Leverandør: VWR Chemicals
Danger
Stabilisering: stabilisert med metanol 9,0- 15,0 % Vol.
87837.180EA 4925 NOK
87837.180
Formaldehyd 35% in aqueous solution stabilised Reag. Ph. Eur. 1039101
Formaldehyd
Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formel: CH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8
FN: 1198
ADR: 3 (8),III