Formamid treated Reag. Ph. Eur. 1039201

Leverandør: VWR Chemicals

Synonymer: Carbamaldehyde

87838.180EA 930 NOK
87838.180
Formamid treated Reag. Ph. Eur. 1039201
Formamid

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: HCONH₂
MW: 45.04 g/mol
Kokepunkt: 210
Smeltepunkt: 2
Flash punkt: 79
MDL nummer: MFCD00007941
CAS-nummer: 75-12-7
Merck Indeks: 13,04261

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR