Indigo carmine solution Reag. Ph. Eur. 1045601

Leverandør: VWR Chemicals

Synonymer: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

87852.290EA 2175 NOK
87852.290
Indigo carmine solution Reag. Ph. Eur. 1045601
Indigo carmine

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0
EINECS: 212-728-8
Merck Indeks: 13,04969

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR