Jod 5 g/l in chloroform Reag. Ph. Eur. 1045805

Leverandør: VWR Chemicals

87855.290EA 3625 NOK
87855.290
Jod 5 g/l in chloroform Reag. Ph. Eur. 1045805
Jod

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: I₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011355
CAS-nummer: 7553-56-2
EINECS: 231-442-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR