Ninhydrin and stannous chloride reagent Reag. Ph. Eur. 1058301
Leverandør: VWR Chemicals
87875.180EA 2600 NOK
87875.180
Ninhydrin and stannous chloride reagent Reag. Ph. Eur. 1058301
Ninhydrin and stannous chloride reagent
2,2% ninhydrin with 0,09% tin II chloride in aqueous solution.
Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.