Kaliumklorid 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101

Leverandør: VWR Chemicals

Synonymer: Potassiumchloride

87898.290EA 1375 NOK
87898.290
Kaliumklorid 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101
Kaliumklorid

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: KCl
MW: 74.55 g/mol
Kokepunkt: 1420
Smeltepunkt: 773
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8
Merck Indeks: 14,07621

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR