Kaliumdihydrogenortofosfat 0.2 mol/l vandig løsning Reag. Ph. Eur. 1069601

Leverandør: VWR Chemicals

87901.290EA 3225 NOK
87901.290
Kaliumdihydrogenortofosfat 0.2 mol/l vandig løsning Reag. Ph. Eur. 1069601
Kaliumdihydrogenortofosfat

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: KH₂PO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011401
CAS-nummer: 7778-77-0
EINECS: 231-913-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR