Amoniakkløsning 25% ≥99.99% (metals basis)

Leverandør: Thermo Fisher Scientific
Danger

87903.AK 87903.AU
87903.AKEA 2150 NOK
87903.AK 87903.AU
Amoniakkløsning 25% ≥99.99% (metals basis)
Amoniakkløsning

Purity: 99.99% CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.91 Appearance: Colourless Odour: Ammonia-like pH: 12 Merck Reference: 14,494 UN No.: 2672 MDL No.: MFCD00066650

Formel: NH₃
Kokepunkt: 32…37,7 °C (1013 hPa)
Tetthet: 0,91
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00066650
CAS-nummer: 1336-21-6
EINECS: 215-647-6
FN: 2672
ADR: 8,III
Merck Indeks: 14,494

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR