Sulphomolybdic reagent R3 Reag. Ph. Eur. 1086500

Leverandør: VWR Chemicals

87947.180EA 870 NOK
87947.180
Sulphomolybdic reagent R3 Reag. Ph. Eur. 1086500
Sulphomolybdic reagents

Starting materials: ammonium molybdate 0.04 mol/l R / water R / sulphuric acid 50% R


Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: (NH₄)₆Mo₇O₂₄·4H₂O
MDL nummer: MFCD00011596
CAS-nummer: 12054-85-2
EINECS: 234-722-4
FN: 2796
ADR: 8 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR