Formaldehyd 0,4% in sulphuric acid 95% Reag. Ph. Eur. 1086805

Leverandør: VWR Chemicals
Danger

87951.180EA 1550 NOK
87951.180
Formaldehyd 0,4% in sulphuric acid 95% Reag. Ph. Eur. 1086805
Formaldehyd

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: CH₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8
FN: 1830
ADR: 8,II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR