Vann Reag. Ph. Eur. 1095503 for chromatography
Leverandør: VWR Chemicals
Synonymer: Hydrogen oxide
87960.290EA 567 NOK
87960.290
Vann Reag. Ph. Eur. 1095503 for chromatography
Vann
Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formel: H₂O
MW: 18.02 g/mol
Kokepunkt: 100
Smeltepunkt: 0
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098