Vann Reag. Ph. Eur. 1095506 nitrate-free

Leverandør: VWR Chemicals

Synonymer: Hydrogen oxide

87961.290EA 600 NOK
87961.290
Vann Reag. Ph. Eur. 1095506 nitrate-free
Vann

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: H₂O
MW: 18.02 g/mol
Kokepunkt: 100
Smeltepunkt: 0
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR