Zinkklorid 20% in formic acid 68% Reag. Ph. Eur. 1096601

Leverandør: VWR Chemicals
Danger

Synonymer: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

87965.290EA 7250 NOK
87965.290
Zinkklorid 20% in formic acid 68% Reag. Ph. Eur. 1096601
Zinkklorid

Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formel: ZnCl₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011295
CAS-nummer: 7646-85-7
EINECS: 231-592-0
FN: 1760
ADR: 8,II

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR