Natrium 1000 mg/L Na in 5% salpetersyre (fra Na₂CO₃) Specpure® standard til AAS

Leverandør: Thermo Fisher Scientific

88097.AE 88097.AP
88097.AEEA 645 NOK
88097.AE 88097.AP
Natrium 1000 mg/L Na in 5% salpetersyre (fra Na₂CO₃) Specpure® standard til AAS
Standarder Kationstandarder Sodium Standards

Molecular Formula: Na₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085307


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR