Terbium 1000 mg/L Tb in 5% salpetersyre (fra Tb₄O₇) Specpure® standard til AAS

Leverandør: Thermo Fisher Scientific

88110.AE 88110.AP
88110.AEEA 695 NOK
88110.AE 88110.AP
Terbium 1000 mg/L Tb in 5% salpetersyre (fra Tb₄O₇) Specpure® standard til AAS
Standarder Kationstandarder Terbium Standards

Molecular formula: Tb₄O₇ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011256


Sertifiseringer: Traceable to NIST.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR