VWR®LABWASH® Classic Universal, Detergent and cleaning agent for ultrasonic baths
Leverandør: VWR Chemicals
88289.360EA 1700 NOK
88289.360
VWR®LABWASH® Classic Universal, Detergent and cleaning agent for ultrasonic baths
Rengjøringsløsninger Detergenter og overflatevasking
Alkaline, powerful universal cleaning agent for use in ultrasonic baths or for manual cleaning in an immersion bath.
  • Powerful cleaning agent
  • High material compatibility
  • Contains special corrosion inhibitors
  • Weakly foaming
VWR LABWASH® Classic Universal is characterised by a high content of phosphates in combination with dispersing aids and surfactants. This combination produces a very strong cleaning result against organic and inorganic soiling, such as oils, proteins, greases, resins etc. Even burnt-on and encrusted structures are dissolved andMore Product Information