Boron 5.000 mg/l B i vandig
Leverandør: VWR Chemicals
88380.260EA 4225 NOK
88380.260 88380.290
Boron 5.000 mg/l B i vandig
Standarder Kationstandarder Boron Standards
This certified reference material is produced using a high-purity starting material, acid from sub-boiling and 18 MOhm deionised water.