Organic reference standard, 8-Methyl-6-nonenoic acid, predominantly trans, analytical standard, Supelco®

Leverandør: Merck

89117-100MG
89117-100MGEA 1850 NOK
89117-100MG
Organic reference standard, 8-Methyl-6-nonenoic acid, predominantly trans, analytical standard, Supelco®
Standarder Organiske standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a16a89283cae0653800a41057f248cb74887d889&vid=a16a89283cae0653800a41057f248cb74887d889&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Beskrivelse 8-Methyl-6-nonenoic acid
Environmentally Preferable
CAS-nr 59320-77-3
Formel C₁₀H₁₈O₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR