Eddiksyre 2 %

Leverandør: VWR Chemicals

Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

89255.290EA 375 NOK
89255.290
Eddiksyre 2 %
Eddiksyre glasial (iseddik)
Formel: H₃CCOOH
MDL nummer: MFCD00036152
CAS-nummer: 64-19-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR