Multi-element Ion chromatography cation standard solution, certified, Supelco®
Leverandør: Merck
89316-50ML-F
89316-50ML-FEA 3400 NOK
89316-50ML-F
Multi-element Ion chromatography cation standard solution, certified, Supelco®
Standarder Kationstandarder Multi-Element Standards
10,0 mg/kg ±0,2% Li+, Na+, K+, Mg²⁺, Ca²⁺ each cation, certified reference material traceable to SI.