Kaliumhydroksid 1 mol/l (1 N) in methanol, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals

Synonymer: Potash caustic, Caustic potash

89346.260EA 1150 NOK
89346.260
Kaliumhydroksid 1 mol/l (1 N) in methanol, AVS TITRINORM volumetric solution
Kaliumhydroksid
Formel: KOH
MDL nummer: MFCD00003553
CAS-nummer: 1310-58-3
FN: 2924
ADR: 3 (8),II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR