Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 99.0-101.0% (by argentometric titration), LiChropur™ derivatisation reagent, for HPLC, Supelco®

Leverandør: Merck

Synonymer: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

89397-10G
89397-10GEA 240 NOK
89397-10G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 99.0-101.0% (by argentometric titration), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Girard's reagent T (carbazoylmethyltrimethylammonium chloride)

LC-MS: suitable for derivatisation.

Formel: C₅H₁₄ClN₃O
MW: 167.64 g/mol
MDL nummer: MFCD00012009
CAS-nummer: 123-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf1675d922dfcc9bcc19e39d752235874c12b735&vid=cf1675d922dfcc9bcc19e39d752235874c12b735&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR