Kaliumklorid 0.2 mol/l (0.2 N) in aqueous solution volumetric solution

Leverandør: VWR Chemicals

Synonymer: Potassiumchloride

89511.290EA 490 NOK
89511.290
Kaliumklorid 0.2 mol/l (0.2 N) in aqueous solution volumetric solution
Kaliumklorid
Formel: KCl
MW: 74.55 g/mol
Kokepunkt: 1420
Smeltepunkt: 773
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
Merck Indeks: 14,07621

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR