p-Toluidin, Supelco®

Leverandør: Merck
Danger


M-Clarity™ quality level = MQ100

89630-100G 89630-500G
89630-100GEA 1260 NOK
89630-100G 89630-500G
p-Toluidin, Supelco®
p-Toluidin
Formel: C₇H₉N
Kokepunkt: 201 °C (1013 hPa)
Smeltepunkt: 43 °C
Tetthet: 1,046 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007906
CAS-nummer: 106-49-0
EINECS: 203-403-1
FN: 3451
ADR: 6.1,II
Merck Indeks: 12,09674
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53e3fb888b42d603a36c18dcef45266025bd53f3&vid=b91c594bab10758721c830a0f7da73fad5c65d63&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR