Organic reference standard, Citrus Red 2, analytical standard, Supelco®
Leverandør: Merck
89774-25MG
89774-25MGEA 1800 NOK
89774-25MG
Organic reference standard, Citrus Red 2, analytical standard, Supelco®
Standarder Organiske standarder
Citrus red 2 is an artificial dye, which belongs to the mono-azo group of dyes.
  • Used to color orange skins
Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31357bcf214a2aa222913eb1de731e889d0668ec&vid=31357bcf214a2aa222913eb1de731e889d0668ec&prid=web&clid=SAPDAM